3,5-Dimethyl-α-(1-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214417-1G 214417-500MG 214417-5G 214417-25G
77466-906EA 415.09 USD
77466-906 77479-638 77489-924 77500-184
3,5-Dimethyl-α-(1-methylpropyl)benzenemethanol ≥95%
3,5-Dimethyl-α-(1-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18871644
CAS Number: 1343621-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR