2-Fluoro-α-(1-methylpropyl)-3-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214397-1G 214397-500MG 214397-5G 214397-25G
77466-902EA 415.09 USD
77466-902 77479-634 77489-920 77500-180
2-Fluoro-α-(1-methylpropyl)-3-(trifluoromethyl)benzenemethanol ≥95%
2-Fluoro-α-(1-methylpropyl)-3-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₄F₄O
MW: 250.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22906654
CAS Number: 1505014-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR