3-Ethoxy-α-(1-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214370-1G 214370-500MG 214370-5G 214370-25G
77466-898EA 260.59 USD
77466-898 77479-630 77489-916 77500-176
3-Ethoxy-α-(1-methylpropyl)benzenemethanol ≥95%
3-Ethoxy-α-(1-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21928910
CAS Number: 1533626-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR