4-(1-Hydroxy-2-methylbutyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

214258-1G 214258-500MG 214258-5G
77466-894EA 929.06 USD
77466-894 77479-626 77489-910
4-(1-Hydroxy-2-methylbutyl)benzonitrile ≥95%
4-(1-Hydroxy-2-methylbutyl)benzonitrile

Precaución: For research use only

Formula: C₁₂H₁₅NO
MW: 189.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24217308
CAS Number: 864266-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR