1-(6-Bromopyridin-3-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

214222-1G 214222-500MG 214222-5G
77466-890EA 519.12 USD
77466-890 77479-622 77489-906
1-(6-Bromopyridin-3-yl)propan-1-one ≥95%
1-(6-Bromopyridin-3-yl)propan-1-one

Precaución: For research use only

Formula: C₈H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD18207831
CAS Number: 1194020-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR