5-Chloro-2-fluoro-3-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

213746-1G 213746-500MG 213746-5G
77466-886EA 696.28 USD
77466-886 77479-618 77489-902
5-Chloro-2-fluoro-3-methylbenzonitrile ≥95%
5-Chloro-2-fluoro-3-methylbenzonitrile

Precaución: For research use only

Formula: C₈H₅ClFN
Storage Temperature: Ambient
MDL Number: MFCD28751907
CAS Number: 1807116-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR