2-Fluoronicotinamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoropyridine-3-carboxamide

21249-1G 21249-500MG 21249-5G 21249-25G
77466-882EA 115.36 USD
77466-882 77479-614 77489-898 77500-166
2-Fluoronicotinamide ≥97%
2-Fluoronicotinamide

Precaución: For research use only

Formula: C₆H₅FN₂O
MW: 140.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100137
CAS Number: 364-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR