2-Bromo-1-isopropoxy-4-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

21102-1G 21102-500MG 21102-5G
77466-878EA 477.92 USD
77466-878 77479-610 77489-894
2-Bromo-1-isopropoxy-4-isopropylbenzene ≥95%
2-Bromo-1-isopropoxy-4-isopropylbenzene

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356554
CAS Number: 1369849-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR