α,3,5-Trimethyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210769-1G 210769-500MG 210769-5G
77466-874EA 648.9 USD
77466-874 77479-606 77489-890
α,3,5-Trimethyl-α-(1-methylethyl)benzenemethanol ≥96%
α,3,5-Trimethyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12767787
CAS Number: 1182749-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR