α-Methyl-α-(1-methylethyl)-3-(methylthio)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210737-1G 210737-500MG 210737-5G 210737-25G
77466-870EA 339.9 USD
77466-870 77479-602 77489-886 77500-156
α-Methyl-α-(1-methylethyl)-3-(methylthio)benzenemethanol ≥96%
α-Methyl-α-(1-methylethyl)-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528562
CAS Number: 1379311-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR