α-Methyl-α,3-bis(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210709-1G 210709-500MG 210709-5G 210709-25G
77466-866EA 397.58 USD
77466-866 77479-598 77489-882 77500-154
α-Methyl-α,3-bis(1-methylethyl)benzenemethanol ≥96%
α-Methyl-α,3-bis(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD20526635
CAS Number: 1443338-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR