4-Methoxy-α,3,5-trimethyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210704-1G 210704-500MG 210704-5G
77466-862EA 648.9 USD
77466-862 77479-594 77489-878
4-Methoxy-α,3,5-trimethyl-α-(1-methylethyl)benzenemethanol ≥96%
4-Methoxy-α,3,5-trimethyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20528536
CAS Number: 1443354-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR