5-Bromo-2-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210698-1G 210698-500MG 210698-5G
77466-858EA 447.02 USD
77466-858 77479-590 77489-874
5-Bromo-2-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%
5-Bromo-2-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23759091
CAS Number: 1511175-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR