3-Bromo-2-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210683-1G 210683-500MG 210683-5G
77466-854EA 447.02 USD
77466-854 77479-586 77489-870
3-Bromo-2-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥96%
3-Bromo-2-fluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867179
CAS Number: 1697866-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR