α-Methyl-α-(1-methylethyl)[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

210660-1G 210660-500MG 210660-5G 210660-25G
77466-850EA 415.09 USD
77466-850 77479-582 77489-866 77500-148
α-Methyl-α-(1-methylethyl)[1,1'-biphenyl]-4-methanol ≥95%
α-Methyl-α-(1-methylethyl)[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₇H₂₀O
MW: 240.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20811150
CAS Number: 860590-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR