3-Bromo-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210654-1G 210654-500MG 210654-5G 210654-25G
77466-846EA 415.09 USD
77466-846 77479-578 77489-862 77500-144
3-Bromo-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%
3-Bromo-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068094
CAS Number: 28114-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR