2-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210649-1G 210649-500MG 210649-5G 210649-25G
77466-842EA 415.09 USD
77466-842 77479-574 77489-858 77500-142
2-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥95%
2-Fluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068019
CAS Number: 1157636-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR