α-Methyl-α-(1-methylethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210633-1G 210633-500MG 210633-5G
77466-838EA 648.9 USD
77466-838 77479-570 77489-854
α-Methyl-α-(1-methylethyl)benzenemethanol ≥96%
α-Methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
MW: 164.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01013588
CAS Number: 4383-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR