α-(1,1-Dimethylethyl)-4-fluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210625-1G 210625-500MG 210625-5G 210625-25G
77466-834EA 415.09 USD
77466-834 77479-566 77489-850 77500-138
α-(1,1-Dimethylethyl)-4-fluoro-α-methylbenzenemethanol ≥97%
α-(1,1-Dimethylethyl)-4-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
Storage Temperature: Ambient
MDL Number: MFCD21328238
CAS Number: 1491375-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR