4-Bromo-α-(1,1-dimethylethyl)-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210623-1G 210623-500MG 210623-5G 210623-25G
77466-830EA 415.09 USD
77466-830 77479-562 77489-846 77500-134
4-Bromo-α-(1,1-dimethylethyl)-α-methylbenzenemethanol ≥95%
4-Bromo-α-(1,1-dimethylethyl)-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD21328245
CAS Number: 1498894-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR