α-(1,1-Dimethylethyl)-2,5-difluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210608-1G 210608-500MG 210608-5G 210608-25G
77466-826EA 297.67 USD
77466-826 77479-558 77489-842 77500-130
α-(1,1-Dimethylethyl)-2,5-difluoro-α-methylbenzenemethanol ≥97%
α-(1,1-Dimethylethyl)-2,5-difluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD24199047
CAS Number: 1547265-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR