α-(1,1-Dimethylethyl)-2,3-difluoro-α-methylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

210579-1G 210579-500MG 210579-5G 210579-25G
77466-822EA 397.58 USD
77466-822 77479-554 77489-838 77500-126
α-(1,1-Dimethylethyl)-2,3-difluoro-α-methylbenzenemethanol ≥96%
α-(1,1-Dimethylethyl)-2,3-difluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD24199060
CAS Number: 1552981-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR