3-Bromo-α-(1,1-dimethylethyl)-2,6-difluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210570-1G 210570-500MG 210570-5G
77466-818EA 447.02 USD
77466-818 77479-550 77489-834
3-Bromo-α-(1,1-dimethylethyl)-2,6-difluoro-α-methylbenzenemethanol ≥97%
3-Bromo-α-(1,1-dimethylethyl)-2,6-difluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₅BrF₂O
MW: 293.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867163
CAS Number: 1696343-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR