α-(1,1-Dimethylethyl)-3-fluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210521-1G 210521-500MG 210521-5G 210521-25G
77466-814EA 415.09 USD
77466-814 77479-546 77489-830 77500-122
α-(1,1-Dimethylethyl)-3-fluoro-α-methylbenzenemethanol ≥97%
α-(1,1-Dimethylethyl)-3-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21328240
CAS Number: 147611-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR