3-Bromo-2,6-difluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210503-1G 210503-500MG 210503-5G
77466-810EA 447.02 USD
77466-810 77479-542 77489-826
3-Bromo-2,6-difluoro-α-methyl-α-(1-methylethyl)benzenemethanol ≥97%
3-Bromo-2,6-difluoro-α-methyl-α-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34867124
CAS Number: 1700665-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR