α-(1,1-Dimethylethyl)-4-methoxy-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210483-1G 210483-500MG 210483-5G 210483-25G
77466-806EA 297.67 USD
77466-806 77479-538 77489-822 77500-116
α-(1,1-Dimethylethyl)-4-methoxy-α-methylbenzenemethanol ≥95%
α-(1,1-Dimethylethyl)-4-methoxy-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD21328250
CAS Number: 147611-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR