α-(1-Ethylpropyl)-5-fluoro-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210475-1G 210475-500MG 210475-5G 210475-25G
77466-802EA 415.09 USD
77466-802 77479-534 77489-818 77500-112
α-(1-Ethylpropyl)-5-fluoro-2-methoxybenzenemethanol ≥95%
α-(1-Ethylpropyl)-5-fluoro-2-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD12787633
CAS Number: 1247744-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR