4-Ethoxy-α-(1-ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210464-1G 210464-500MG 210464-5G 210464-25G
77466-798EA 415.09 USD
77466-798 77479-530 77489-814 77500-108
4-Ethoxy-α-(1-ethylpropyl)benzenemethanol ≥95%
4-Ethoxy-α-(1-ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786980
CAS Number: 1249482-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR