α-(1-Ethylpropyl)-2,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210460-1G 210460-500MG 210460-5G 210460-25G
77466-794EA 297.67 USD
77466-794 77479-526 77489-810 77500-104
α-(1-Ethylpropyl)-2,5-dimethylbenzenemethanol ≥95%
α-(1-Ethylpropyl)-2,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD12787267
CAS Number: 1250398-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR