α-(1-Ethylpropyl)[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

210447-1G 210447-500MG 210447-5G 210447-25G
77466-790EA 415.09 USD
77466-790 77479-522 77489-806 77500-100
α-(1-Ethylpropyl)[1,1'-biphenyl]-4-methanol ≥95%
α-(1-Ethylpropyl)[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₈H₂₂O
MW: 254.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19486804
CAS Number: 1283328-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR