α-(1-Ethylpropyl)-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210436-1G 210436-500MG 210436-5G 210436-25G
77466-786EA 364.62 USD
77466-786 77479-518 77489-802 77500-096
α-(1-Ethylpropyl)-2-methylbenzenemethanol ≥95%
α-(1-Ethylpropyl)-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD19601806
CAS Number: 1339593-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR