α-(1-Ethylpropyl)-3-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210432-1G 210432-500MG 210432-5G 210432-25G
77466-782EA 353.29 USD
77466-782 77479-514 77489-798 77500-092
α-(1-Ethylpropyl)-3-methoxybenzenemethanol ≥95%
α-(1-Ethylpropyl)-3-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19601772
CAS Number: 1340056-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR