α-(1-Ethylpropyl)-2-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210426-1G 210426-500MG 210426-5G 210426-25G
77466-778EA 297.67 USD
77466-778 77479-510 77489-794 77500-088
α-(1-Ethylpropyl)-2-fluorobenzenemethanol ≥97%
α-(1-Ethylpropyl)-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19601802
CAS Number: 1341921-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR