α-(1-Ethylpropyl)-4-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210414-1G 210414-500MG 210414-5G 210414-25G
77466-774EA 415.09 USD
77466-774 77479-506 77489-790 77500-084
α-(1-Ethylpropyl)-4-(2-methylpropyl)benzenemethanol ≥95%
α-(1-Ethylpropyl)-4-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₆H₂₆O
MW: 234.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18575455
CAS Number: 1406297-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR