α-(1-Ethylpropyl)-3-(1-methylethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210393-1G 210393-500MG 210393-5G
77466-770EA 447.02 USD
77466-770 77479-502 77489-786
α-(1-Ethylpropyl)-3-(1-methylethoxy)benzenemethanol ≥95%
α-(1-Ethylpropyl)-3-(1-methylethoxy)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23384822
CAS Number: 1505675-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR