2-Chloro-α-(1-ethylpropyl)-6-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210376-1G 210376-500MG 210376-5G 210376-25G
77466-766EA 297.67 USD
77466-766 77479-498 77489-782 77500-078
2-Chloro-α-(1-ethylpropyl)-6-fluorobenzenemethanol ≥95%
2-Chloro-α-(1-ethylpropyl)-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD23833755
CAS Number: 1516765-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR