3-Bromo-α-(1-ethylpropyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210360-1G 210360-500MG 210360-5G 210360-25G
77466-762EA 297.67 USD
77466-762 77479-494 77489-778 77500-074
3-Bromo-α-(1-ethylpropyl)benzenemethanol ≥97%
3-Bromo-α-(1-ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23833756
CAS Number: 1540543-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR