α-(1-Ethylpropyl)-3-fluoro-5-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210352-1G 210352-500MG 210352-5G
77466-758EA 497.49 USD
77466-758 77479-490 77489-774
α-(1-Ethylpropyl)-3-fluoro-5-(trifluoromethyl)benzenemethanol ≥95%
α-(1-Ethylpropyl)-3-fluoro-5-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₆F₄O
MW: 264.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25680610
CAS Number: 1545452-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR