α-(1-Ethylpropyl)-2,6-difluoro-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210348-1G 210348-500MG 210348-5G 210348-25G
77466-754EA 415.09 USD
77466-754 77479-486 77489-770 77500-068
α-(1-Ethylpropyl)-2,6-difluoro-3-methylbenzenemethanol ≥95%
α-(1-Ethylpropyl)-2,6-difluoro-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310354
CAS Number: 1551632-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR