2-Bromo-α-(1-ethylpropyl)-6-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210342-1G 210342-500MG 210342-5G 210342-25G
77466-750EA 226.6 USD
77466-750 77479-480 77489-764 77500-062
2-Bromo-α-(1-ethylpropyl)-6-fluorobenzenemethanol ≥97%
2-Bromo-α-(1-ethylpropyl)-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD34867103
CAS Number: 1592398-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR