α-(1-Ethylpropyl)-3-methoxy-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210329-1G 210329-500MG 210329-5G 210329-25G
77466-746EA 350.2 USD
77466-746 77479-478 77489-762 77500-060
α-(1-Ethylpropyl)-3-methoxy-4-methylbenzenemethanol ≥95%
α-(1-Ethylpropyl)-3-methoxy-4-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867091
CAS Number: 1596647-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR