α-(1-Ethylpropyl)-4-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210320-1G 210320-500MG 210320-5G 210320-25G
77466-742EA 397.58 USD
77466-742 77479-474 77489-758 77500-056
α-(1-Ethylpropyl)-4-(methylthio)benzenemethanol ≥95%
α-(1-Ethylpropyl)-4-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867082
CAS Number: 1598041-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR