4-Bromo-α-(1-ethylpropyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

210260-1G 210260-500MG 210260-5G 210260-25G
77466-738EA 297.67 USD
77466-738 77479-470 77489-754 77500-054
4-Bromo-α-(1-ethylpropyl)benzenemethanol ≥97%
4-Bromo-α-(1-ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786809
CAS Number: 1249709-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR