α-(1-Ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210253-1G 210253-500MG 210253-5G 210253-25G
77466-734EA 297.67 USD
77466-734 77479-466 77489-750 77500-050
α-(1-Ethylpropyl)benzenemethanol ≥95%
α-(1-Ethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786905
CAS Number: 854179-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR