1-(3-Ethyl-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

209976-1G 209976-500MG 209976-5G
77466-730EA 696.28 USD
77466-730 77479-462 77489-744
1-(3-Ethyl-5-methylphenyl)ethanone ≥95%
1-(3-Ethyl-5-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867019
CAS Number: 2384172-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR