1-(3-Isopropylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-[3-(1-Methylethyl)phenyl]ethanone

209947-1G 209947-500MG 209947-5G 209947-25G
77466-722EA 105.06 USD
77466-722 77479-454 77489-736 77500-040
1-(3-Isopropylphenyl)ethanone ≥95%
1-(3-Isopropylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134684
CAS Number: 40428-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR