3-Iodo-1,2,4,5-tetrafluorobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Iodo-4H-tetrafluorobenzene, 1,2,4,5-tetrafluoro-3-iodobenzene, 1,2,4,5-Tetrafluoro-3-iodobenzene, 4-Iodo-1H-tetrafluorobenzene

209769-1G 209769-500MG 209769-5G 209769-25G
77466-718EA 237.93 USD
77466-718 77479-450 77489-730 77500-034
3-Iodo-1,2,4,5-tetrafluorobenzene ≥95%
3-Iodo-1,2,4,5-tetrafluorobenzene

Precaución: For research use only

Formula: C₆HF₄I
MW: 275.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09763612
CAS Number: 5243-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR