2-(Pentafluorophenyl)-2-propanol ≥95%

Supplier: AOBChem USA

Warning

209738-1G 209738-500MG 209738-5G 209738-25G
77466-714EA 91.67 USD
77466-714 77479-446 77489-726 77500-030
2-(Pentafluorophenyl)-2-propanol ≥95%
2-(Pentafluorophenyl)-2-propanol

Precaución: For research use only

Formula: C₉H₇F₅O
Storage Temperature: Ambient
MDL Number: MFCD00039613
CAS Number: 715-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR