2,3,5,6-Tetrafluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

208824-1G 208824-500MG 208824-5G 208824-25G
77466-710EA 295.61 USD
77466-710 77479-442 77489-718 77500-020
2,3,5,6-Tetrafluorobenzamide ≥97%
2,3,5,6-Tetrafluorobenzamide

Precaución: For research use only

Formula: C₇H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD00456801
CAS Number: 28293-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR