α-Cyclohexyl-4-(1,1-dimethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

208449-1G 208449-500MG 208449-5G 208449-25G
77466-706EA 415.09 USD
77466-706 77479-438 77489-712 77500-014
α-Cyclohexyl-4-(1,1-dimethylpropyl)benzenemethanol ≥95%
α-Cyclohexyl-4-(1,1-dimethylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₈H₂₈O
Storage Temperature: Ambient
MDL Number: MFCD23209364
CAS Number: 1537699-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR